Temperature chart recorders, SK4 and SL4
Supplier: DICKSON
Certificates
About this item
These self-contained temperature recorders on a Ø 100 mm disc provide a permanent record of temperatures with an average response time of 11 minutes to 63% of full scale. Powered by a AA battery, they are used for monitoring and preparing stocks of vaccines, foods and pharmaceutical products. SL4 models include an LCD screen for displaying data (W×D: 48×16 mm).
- Long term recording during storage, transport or shipping
- Hinged door
- Commuter for recording 7 days or 24 hours
W×D×H: 132×132×61 mm
Weight: 295 g
Charts
Product Information
Frequently Bought Together