Order Entry
United Kingdom
ContactUsLinkComponent
Medical wipes, Kleenex
Medical wipes, Kleenex
Catalog # KIMB7432
Supplier:  KIMBERLY CLARK
undefined
Medical wipes, Kleenex
Catalog # KIMB7432
Supplier:  KIMBERLY CLARK

Specifications

  • Description:
    Medical wipes, 2 ply
  • L×W:
    186×108 mm
  • Environmentally Preferable:
  • Pk:
    80

Specifications

About this item

Kleenex® Wipes are ideal in busy healthcare environments. Available in a handy, portable POP-UP box, these wipes are soft, yet strong and can be used for multiple tasks.

  • Ideal in busy healthcare environments
  • Wipes are soft, yet strong and can be used for multiple tasks
  • Portable POP-UP Box

Individual sheet size 18.6 cm (L) × 10.8 cm (W).

Certifications: These wipes are FSC certified, giving assurance that they are made from or contain woodfibre that comes from FSC certified forests. These forests are managed to meet the social, economic and ecological needs of present and future generations.