Order Entry
United Kingdom
ContactUsLinkComponent
Cryoware labels, Nalgene®
Cryoware labels, Nalgene®
Catalog # 479-3291
   Low Manufacturing Impact
undefined
Cryoware labels, Nalgene®
Catalog # 479-3291
   Low Manufacturing Impact
Specifications
  • Printer model:
  • Printing method:
  • Colour:
    White
  • Description:
    Cryogenic dots
  • Dimensions:
    25×50 mm
  • Material:
    Cloth
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    200
Specifications
About this item
Cloth labels for use at ultra-low temperatures, ballpoint pen compatible.
  • Will not shrink
Dimensions: 25×50 mm.
Delivery: 20 labels/sheet, 10 sheets/pack.