Cryogenic tubes, CryoTubes™, Nunc™
About this item
PP tubes with PP screw cap (internal thread vials) or PE screw cap (external thread vials) and white writing area, sterile. Developed for storage of cell cultures, bacteria and other samples at extremely low temperatures. Suitable for use in freezers as well as in the vapour phase of liquid nitrogen*.
Non toxic, non pyrogenic.
Conform to IATA regulations for the air transportation of biological samples.
* Warning: Do not use CryoTubes™ in the liquid phase of liquid nitrogen unless correctly sealed in Nunc CryoFlex™ tubing. Improper use may cause entrapment of liquefied nitrogen inside the vial and lead to pressure build up, resulting in possible explosion or biohazard release. The tubes 479-6816 and 479-6853 are suitable for storage to −80 °C only.
* without writing area
** free standing
*** starfoot
18 Options Available Below
Product Details & Documents
PP tubes with PP screw cap (internal thread vials) or PE screw cap (external thread vials) and white writing area, sterile. Developed for storage of cell cultures, bacteria and other samples at extremely low temperatures. Suitable for use in freezers as well as in the vapour phase of liquid nitrogen*.
Non toxic, non pyrogenic.
Conform to IATA regulations for the air transportation of biological samples.
* Warning: Do not use CryoTubes™ in the liquid phase of liquid nitrogen unless correctly sealed in Nunc CryoFlex™ tubing. Improper use may cause entrapment of liquefied nitrogen inside the vial and lead to pressure build up, resulting in possible explosion or biohazard release. The tubes 479-6816 and 479-6853 are suitable for storage to −80 °C only.
* without writing area
** free standing
*** starfoot
Specifications for Product Family
Specifications
- Catalog No:
- 479-6840
- 479-6842
- 479-6837
- 479-6841
- 479-6843
- 479-6844
- 479-6838
- 479-6839
- 479-3202
- 479-6845
- 479-6852
- 479-1159
- 479-6849
- 479-6846
- 479-6847
- 479-6850
- 479-6848
- 479-6836
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- IRECS
- IRECS
- IRECS
Specifications
Product Family Options
Internal thread with silicone O-ring, sterile
Product Information
- CapacityBaseØ×HCap includedCap colourAvailabilityPrice
- 479-6840Cryogenic tubes with writing areaCryogenic tubes with writing area1,00 mlV12,5×42 ** mmYesClearLog in for availabilityLog in for availability1 * 500 items1 * 500 items - £206.00£206.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 500 items£206.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.479-6840Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 479-6842Cryogenic tubes with writing areaCryogenic tubes with writing area1,00 mlV12,5×42 *** mmYesClearLog in for availabilityLog in for availability1 * 500 items1 * 500 items - £209.00£209.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 500 items£209.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.479-6842Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 479-6837Cryogenic tubes with writing areaCryogenic tubes with writing area1,80 mlU12,5×48 mmYesClearLog in for availabilityLog in for availability1 * 500 items1 * 500 items - £184.00£184.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 500 items£184.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.479-6837Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 479-6841Cryogenic tubes with writing areaCryogenic tubes with writing area1,80 mlU12,5×49 ** mmYesClearLog in for availabilityLog in for availability1 * 450 items1 * 450 items - £172.00£172.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 450 items£172.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.479-6841Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 479-6843Cryogenic tubes with writing areaCryogenic tubes with writing area1,80 mlU12,5×49 *** mmYesClearLog in for availabilityLog in for availability1 * 450 items1 * 450 items - £168.00£168.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 450 items£168.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.479-6843Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 479-6844Cryogenic tubes with writing areaCryogenic tubes with writing area3,60 mlU12,5×72 *** mmYesClearLog in for availabilityLog in for availability1 * 400 items1 * 400 items - £185.00£185.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 400 items£185.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.479-6844Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 479-6838Cryogenic tubes with writing areaCryogenic tubes with writing area3,60 mlU12,5×70 mmYesClearLog in for availabilityLog in for availability1 * 400 items1 * 400 items - £180.00£180.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 400 items£180.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.479-6838Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 479-6839Cryogenic tubes with writing areaCryogenic tubes with writing area4,50 mlU12,5×92 mmYesClearLog in for availabilityLog in for availability1 * 300 items1 * 300 items - £131.00£131.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 300 items£131.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.479-6839Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 479-3202Cryogenic tubes with writing areaCryogenic tubes with writing area4,50 mlU12,5×92 mmYesClearLog in for availability1 * 1.200 items£823.00Specifications:
More
Specifications
Cat. No.479-3202Environmentally PreferableEnvironmental certification - 479-6845Cryogenic tubes with writing areaCryogenic tubes with writing area4,50 mlU12,5×92 *** mmYesClearLog in for availabilityLog in for availability1 * 300 items1 * 300 items - £143.00£143.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 300 items£143.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.479-6845Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact
Internal thread with silicone O-ring, without caps, non sterile
Product Information
- CapacityBaseØ×HCap includedCap colourAvailabilityPrice
- 479-6852Cryogenic tubes with writing areaCryogenic tubes with writing area1,80 mlU12,5×48 mmNo—Log in for availabilityLog in for availability1 * 2.500 items1 * 2.500 items - £644.00£644.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 2.500 items£644.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.479-6852Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 479-1159Cryogenic tubes with writing areaCryogenic tubes with writing area3,60 mlU12,5×70 mmNo—Log in for availabilityLog in for availability1 * 1.500 items1 * 1.500 items - £444.00£444.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 1.500 items£444.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.479-1159Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact
External thread, sterile
Product Information
- CapacityBaseØ×HCap includedCap colourAvailabilityPrice
- 479-6849Cryogenic tubesCryogenic tubes1,00 mlV12,5×41 * *** mmYesClearLog in for availabilityLog in for availability1 * 500 items1 * 500 items - £178.00£178.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 500 items£178.00Specifications:More
Specifications
Cat. No.479-6849Environmentally PreferableEnvironmental certification - 479-6846Cryogenic tubes with writing areaCryogenic tubes with writing area1,00 mlV12,5×41 *** mmYesClearLog in for availabilityLog in for availability1 * 500 items1 * 500 items - £192.00£192.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 500 items£192.00Specifications:More
Specifications
Cat. No.479-6846Environmentally PreferableEnvironmental certification - 479-6847Cryogenic tubes with writing areaCryogenic tubes with writing area1,80 mlU12,5×48 *** mmYesClearLog in for availabilityLog in for availability1 * 450 items1 * 450 items - £154.00£154.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 450 items£154.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.479-6847Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECSLow Manufacturing Impact - 479-6850Cryogenic tubesCryogenic tubes1,80 mlU12,5×48 * *** mmYesClearLog in for availabilityLog in for availability1 * 450 items1 * 450 items - £156.00£156.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 450 items£156.00Specifications:More
Specifications
Cat. No.479-6850Environmentally PreferableEnvironmental certification - 479-6848Cryogenic tubes with writing areaCryogenic tubes with writing area4,50 mlU12,5×91 *** mmYesClearLog in for availabilityLog in for availability1 * 300 items1 * 300 items - £125.00£125.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 300 items£125.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.479-6848Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECSLow Manufacturing Impact - 479-6836Cryogenic tubesCryogenic tubes4,50 mlU12,5×91 * *** mmYesClearLog in for availabilityLog in for availability1 * 300 items1 * 300 items - £119.00£119.00
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 300 items£119.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.479-6836Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECSLow Manufacturing Impact