Tubing, rubber, vacuum
Supplier: DEUTSCH NEUMANN
Certificates
About this item
Natural rubber, red.
- Flexible, excellent tensile strength
- Economical
Heavy wall, suitable for vacuum applications in the laboratory.
Temperature -resistance: −10 to +50 °C.
Hardness (Shore durometer A): 50.
Frequently Bought Together