Individual PCR tubes with attached caps
: Brand
PP. These 0,2 and 0,5 ml PCR tubes with attached caps are compatible with leading thermal cyclers with heated lids.
- Unique cap design closes and opens easily without tools, yet ensures a tight fit to reduce sample evaporation
- Flat caps allow simple labelling
- Free from DNase, RNase and DNA
Colour: Clear
Frequently Bought Together