Specifications
- Description:Multicassette 6 drug test for urine (AMP / BZO / COC / MAMP / OPI / THC)
- Environmentally Preferable:
- Pk:25
Specifications
About this item
The multicassette tests for up to 12 several drugs simultaneously, and is very easy to use. The urine sample is added to the sample well with the pipette that comes with the kit, and resuts are read at 3 minutes. Tests are individually wrapped in a foil pouch, to ensure the high-quality of the product is retained throughout the shelf-life.
- Test for up to 12 drug types simultaneously
- Easy to perform test routine
- Clear results within 3 minutes
- Greater than 99% accuracy
- Wide range of test combinations available
- Pipette provided for easy sampling
- Detects recent drug use, approx. 1 to 8 days
Recommended markets and use:
On-site screening
Safety critical situations
Random screening
For cause screening
Monitoring rehabilitation
Drug Option Types:
AMP - Amphetamines
BAR - Barbiturates
BUP - Buprenorphine
BZO - Benzodiazepines
COC - Cocaine
EDDP - Methadone Metabolite
FEN - Fentanyl
KET - Ketamine
MAMP - Methamphetamines
MDMA - Ecstasy
MOP - Morphine
MTD - Methadone
OPI - Opiates
OXY - Oxycodone
PCP - Phencyclidine
PPX - Propoxyphene
TCA - Tricyclic Antidepressants
TRA - Tramadol
THC - Cannabis