Bench protectors, Benchkote®, Whatman™
Cytiva's Whatman™ Benchkote® surface protector is an absorbent lab paper that protects surfaces against hazardous spills. After use, the bench protector sheet can be incinerated or disposed of according to local regulations.
- Material is strong and tear resistant when wet or dry
- Smooth white surface can be written on with ink or pencil and lies flat
- Suitable for saturation with disinfectant to protect benches where pathogens and other bacteria are present
- Can be incinerated after use: PE layer is rapidly consumed when incinerated and does not melt or drip
Benchkote® material has a high quality, smooth, absorbent lab paper layer that soaks up liquid spills. A second laminated polyethylene layer prevents flow-through to the working surface.
Frequently Bought Together