Scalpels, ceramic
Supplier: SLG Kunststoff
Certificates
About this item
Particularly high quality and durable hard ceramic material for long lasting, excellent sharpness. No metal parts, thus also suitable for trace analysis. Super-lightweight, sturdy, ergonomic handle for safe and easy handling.
- Anti-magnetic, anti-static, corrosion-free, dishwasher safe, chemically resistant to aggressive media
- Extremely heat resistant, low thermal conductivity
- Regrinding is not necessary
Frequently Bought Together