Pipette tips, Finntip™ Stepper
Supplier: Thermo Fisher Scientific
Certificates
About this item
Full range of positive displacement syringe tips designed for the Finnpipette™ Stepper.
- Ergonomic design
- Seven syringe sizes covering a range of dispensing volumes from 10 µl to 50 ml
- Adapted for applications using aggressive and viscous liquids
Specifications
- Catalog No:
- 613-2641
- 613-2640
- 613-2639
- 613-2638
- 613-2637
- 613-2636
- 613-2647
- 613-2635
- 613-2646
- 613-2634
- 613-2645
- 613-2644
- 613-2643
- 613-2642
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)