Microtitre plates, 96-well
Supplier: Thermo Fisher Scientific
Certificates
About this item
Rigid styrene or flexible vinyl, square well. Ideal for micro-volume applications, such as storage, sample or reagent transfer, serial dilution, haemogglutination, blood typing and compliment fixation. Available with a choice of flat, U- or V-bottom well shape.
- U-bottom well shape improves washing in ELISAs, enhances sensitivity in fluorescent applications and facilitates observation of agglutination reactions
- V-bottom well shape is optimal for magnetic bead assays, ideal for agglutination tests, useful for centrifugation or concentration and facilitates recovery of contents
Specifications
- Catalog No:
- 735-0180
- 735-0194
- 735-0184
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement