Battery acid tester
614-0551
: GERING
- Description:Hydrometer with cylindrical bulb
- Environmentally Preferable:
- Pk:1
- Range:1,1 – 1,3
Battery acid tester with glass cylinder and intake rubber bulb.
: Complete in cardboard case.
Battery acid tester with glass cylinder and intake rubber bulb.