Tissue processing/embedding cassettes, Histosette® II, M485
Supplier: Simport Scientific
Certificates
About this item
Acetal polymer, similar to M492, but covers are already removed from cassettes and are packed separately in the case.
- Convenient for cassette labelling instruments as the covers are already removed
- Resistant to the chemical action of histological solvents
- Flow-through slots maximise fluid exchange
Frequently Bought Together