Urine and biological specimen containers, with screw caps
Supplier: KARTELL
Certificates
About this item
PS, transparent with PE or PP screw caps.
- 60 ml with PE screw caps and 200 ml with PP screw caps
- Quick opening and closing twist cap, leakproof
Ideal for collecting, storing and transporting urine samples and biological specimens.
* Aseptic containers individually wrapped, with label.