Order Entry
United Kingdom
ContactUsLinkComponent
HPLC columns, Develosil® ODS-UG
HPLC columns, Develosil® ODS-UG
Supplier:  HICHROM
Certificates
HPLC columns, Develosil® ODS-UG
Supplier:  HICHROM
Specifications
  • Catalog No:
  • HISMUG11546250W
  • 554-7681
  • Carbon load:
  • 18%
  • 18%
  • Endcapped:
  • Yes
  • Yes
  • Surface area:
  • 300 m²/g
  • 300 m²/g