Spray bottles, LaboPlast®
Supplier: BURKLE
Certificates
About this item
LDPE, translucent, PP spray attachment.
- Good chemical resistance, good temperature resistance
- Spray nozzle adjustment can be varied by rotation
- Pressureless spray bottle
- Media-contacting parts PP, PE and stainless steel
Spray, choice of fine mist or focused jet spray functions to disperse fluids.
Frequently Bought Together