Measuring cylinders, bell-shaped, soda-lime glass
AR glass, clear.
- Very good chemical resistance
- Good thermal resistance
- Fill volume is easy to read
With a broad base for stability. Fused blue graduations, very easy to read. Class B tolerance, twice the official tolerance. No handle.
Frequently Bought Together