Methyl cellulose, 15 cP (2% H₂O), BioReagent cell culture reagent, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Cellulose methyl ether
CAS Number:9004-67-5
:
MDL Number:MFCD00081763
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together