Stainless Steel Trays
Supplier: Adelphi Pharma Hygiene Products
Certificates
About this item
A range of stainless steel trays which meet international pharma and food quality compliance regulations.
- Trays available as standard, off-the-shelf for delivery within days
- 304 food grade stainless steel
- Shallow and deep designs available
- Can be cleaned with alcohol-based disinfectants, and in industrial dishwashers/autoclaves
Available in 304 grade stainless steel, these trays are supplied in one piece, with radiused corners. They are crevice-free and pit-free and are bright polished throughout.
A range of optional modifications are available to meet every requirement, including bespoke etching of logos and other information. Trays are available in Shallow or Deep formats.
Frequently Bought Together