IPTG (isopropyl-β-D-thiogalactopyranoside)
Supplier: BACHEM BIOCHEMICA
Certificates
About this item
Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside
CAS Number:367-93-1
:
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273
Boiling Pt:402
Molecular Weight:238.31 g/mol
Melting Pt:109.5…110.5 °C
Storage Temperature:Freezer
EINECS:206-703-0
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together