Phenolphthalein 0,1% in ethanol 80% Reag. Ph. Eur. 1063702
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
CAS Number:77-09-8
:
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913
Molecular Weight:318.33 g/mol
Merck Index:14,07243
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together