Order Entry
United Kingdom
ContactUsLinkComponent
Vanadyl sulphate pentahydrate 17-23% (v/v)
Vanadyl sulphate pentahydrate 17-23% (v/v)
Certificates
undefined
Vanadyl sulphate pentahydrate 17-23% (v/v)
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    Blue Moist crystalline powder or crystals
  • Degree of hydration:
    ca. 5 moles water
  • Titration with KMnO4:
    17 to 23 % (V)
  • Iron (Fe):
    ≤600 ppm
  • Potassium (K):
    ≤500 ppm
  • Sodium (Na):
    ≤500 ppm