Order Entry
United Kingdom
ContactUsLinkComponent
Anti-SFRS8 Rabbit Polyclonal Antibody
Anti-SFRS8 Rabbit Polyclonal Antibody
  ANTIA100771-100
New Product
undefined
Anti-SFRS8 Rabbit Polyclonal Antibody
  ANTIA100771-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Splicing factor arginine/serine rich 8 (suppressor of white apricot Drosophila homolog)
  • Antigen symbol:
    SFRS8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q12872
  • Antigen synonyms:
    6330437E22Rik|Splicing factor arginine/serine rich 8|Sfrs8 protein|SFRS 8|AW212079|MGC167082|1190005N23Rik|SFRS8|AI197402
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    104 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human SFRS8 (amino acids 201 - 250)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal antibody to SFRS8 for WB and ELISA with samples derived from human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: SFRS8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human