Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-Flt3 ligand Rabbit Polyclonal Antibody
  ANTIA100569-100
New Product
undefined
Anti-Flt3 ligand Rabbit Polyclonal Antibody
  ANTIA100569-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    FL
  • Antigen symbol:
    Flt3 ligand
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P49771
  • Antigen synonyms:
    Flt 3L|Flt3L|FLT3L_HUMAN|Fms related tyrosine kinase 3 ligand|FLT3LG|FLT3 LG|Flt 3 ligand|Flt3 ligand|Flt3 L
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    26 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from the C-terminal region of human FLT3LG (amino acids 171 - 220)
  • Purification:
    The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal antibody to FLT3LG for WB and ELISA with samples derived from human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: Flt3 ligand
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human