Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-Apolipoprotein CII / ApoC-II Rabbit Polyclonal Antibody
  ANTIA8964-100
New Product
undefined
Anti-Apolipoprotein CII / ApoC-II Rabbit Polyclonal Antibody
  ANTIA8964-100
New Product

 

  • Antibody type:
    Primary
  • Antigen symbol:
    Apolipoprotein CII / ApoC-II
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P02655
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
  • Molecular weight:
    11 kDa
  • Sequence:
    MGTRLLPALFLVLLVLGFEVQGTQQPQQDEMPSPTFLTQVKESLSSYWESAKTAAQNLYEKTYLPAVDEKLRDLYSKSTAAMSTYTGIFTDQVLSVLKGEE
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-101 of human APOC2 (NP_000474.2)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to Apolipoprotein CII / ApoC-II for WB with samples derived from Human and Rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:100 to 1:50

Type: Primary
Antigen: Apolipoprotein CII / ApoC-II
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat