Anti-Decorin Mouse Monoclonal Antibody [clone: DCN/6289]
Catalog # ANTIA277573-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Bone proteoglycan II
- Antigen symbol:Decorin
- Clonality:Monoclonal
- Clone:DCN/6289
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Pig,Rabbit,Horse,Elk,Chicken,Human,Bovine,Dog,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P07585
- Antigen synonyms:Decorin|Decorin proteoglycan|DSPG 2|Dermatan sulphate proteoglycans II|DSPG2|CSCD|DCN|DKFZp686J19238|DCN protein
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:43 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment corresponding to the C terminal core of bovine DCN.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [DCN/6289] antibody to Decorin for Flow Cytometry, IF, WB and IHC-P with samples derived from human, bovine, canine, chicken, elk, equine, mouse, porcine and rabbit.
- Validated Applications: Flow Cytometry, IF, WB, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Decorin
Clonality: Monoclonal
Clone: DCN/6289
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Bovine;Dog;Chicken;Elk;Horse;Mouse;Pig;Rabbit