Anti-CD68 Mouse Monoclonal Antibody [clone: KP1]
Catalog # ANTIA253937-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD 68
- Antigen symbol:CD68
- Clonality:Monoclonal
- Clone:KP1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Rabbit,Monkey,Human,Rat,Mouse,Cat
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P34810
- Antigen synonyms:DKFZp686M18236|Gp110|CD68 antigen|CD68|CD68 molecule|CD68_HUMAN|LAMP4|MACROPHAGE ANTIGEN CD68|gp11
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:110 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Subcellular fraction of human alveolar macrophages.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [KP1] antibody to CD68 for IHC-P with samples derived from human,monkey, mouse, rat, rabbit and feline.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: KP1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Monkey;Mouse;Rat;Rabbit;Cat