Anti-Taf6 Rabbit Polyclonal Antibody
Catalog # ANTIA271-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:TAF6 RNA polymerase II
- Antigen symbol:Taf6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:S. cerevisiae
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:E 70/85kD|TAFII80|TAFII 70|TATA box binding protein (TBP) associated factor 80 kD|TAF 6|TAF6|TAF2E|TATA box binding protein (TBP) associated factor RNA polymerase II E 70/85kD|, TATA box binding protein associated factor|TATA box binding protein (TBP) associated factor, RNA polymerase II|80kDa
- Storage buffer:Supplied as antiserum with 0,1% sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant Saccharomyces cerevisiae Taf6 protein (a.a. 1-200, His tag)
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to Taf6 for WB with samples derived from Saccharomyces cerevisiae.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:1000
Type: Primary
Antigen: Taf6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Saccharomyces cerevisiae