Anti-Spt3p Rabbit Polyclonal Antibody
Catalog # ANTIA267-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Positive regulator of Ty transcription
- Antigen symbol:Spt3p
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:S. cerevisiae
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P06844
- Antigen synonyms:Transcription initiation protein SPT3 homolog|SUPT3H|SPT3 like protein|YDR392W|Spt3p|SPT3
- Storage buffer:Supplied as antiserum with 0,1% sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant Saccharomyces cerevisiae Spt protein (a.a. 1-200, His tag)
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to Spt3p for WB and ELISA with samples derived from Saccharomyces cerevisiae.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:2000 to 1:5000
Type: Primary
Antigen: Spt3p
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Saccharomyces cerevisiae