Order Entry
United Kingdom
ContactUsLinkComponent
Anti-MCP2 Mouse Monoclonal Antibody [Clone: CCL8/3312]
Anti-MCP2 Mouse Monoclonal Antibody [Clone: CCL8/3312]
Catalog # ANTIA277786-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-MCP2 Mouse Monoclonal Antibody [Clone: CCL8/3312]
Catalog # ANTIA277786-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C C motif chemokine 8
  • Antigen symbol:
    MCP2
  • Clonality:
    Monoclonal
  • Clone:
    CCL8/3312
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P80075
  • Antigen synonyms:
    HC14|Chemokine (C C motif) ligand 8|MCP 2|MCP-2|CCL 8|Chemokine ligand 8|MCP-2(6-76)|Ccl8|CCL8_HUMAN
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    11 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 24-98, of human MCP2 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [CCL8/3312] antibody to MCP2 for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: MCP2
Clonality: Monoclonal
Clone: CCL8/3312
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human