Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Ataxin 1 Rabbit Polyclonal Antibody
Anti-Ataxin 1 Rabbit Polyclonal Antibody
  ANTIA96343-100
New Product
undefined
Anti-Ataxin 1 Rabbit Polyclonal Antibody
  ANTIA96343-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    alternative ataxin1
  • Antigen symbol:
    Ataxin 1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P54253
  • Antigen synonyms:
    D6S504E|Ataxin 1|Spinocerebellar ataxia type 1 protein|ATX1_HUMAN|Ataxin-1|SCA1|ATX1|Atxn1|OTTHUMP00000016065
  • Storage buffer:
    Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
  • Molecular weight:
    87 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human Ataxin 1 (amino acids 742-791)
  • Tested applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal antibody to Ataxin 1 for IHC, IF and ELISA with samples derived from Human and Mouse.

  • Validated applications: IHC, IF, ELISA
  • Recommended dilutions: IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:5000

Type: Primary
Antigen: Ataxin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse