Anti-Peripherin Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM
New Product
Certificates
About this item
Rabbit polyclonal antibody to Peripherin for WB, ICC/IF and IHC with samples derived from Human, Rat, Mouse, Porcine and Bovine.
- Validated applications: WB, ICC/IF, IHC
- Recommended dilutions: WB: 1:1000 to 1:2000, ICC/IF: 1:2000, IHC: 1:200
Type: Primary
Antigen: Peripherin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse;Porcine;Bovine
Specifications
- Catalog No:
- ANTIA85430-50
- ANTIA85430-100
- Antigen symbol:
- Peripherin
- Peripherin
- Antigen name:
- NEF 4
- NEF 4
- Antigen synonyms:
- prph|Neurofilament 4 (57kD)|Neurofilament 4|NEF4|PRPH 1|PRPH1|Peripherin|PERI_HUMAN|Perf
- prph|Neurofilament 4 (57kD)|Neurofilament 4|NEF4|PRPH 1|PRPH1|Peripherin|PERI_HUMAN|Perf
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Pig, Human, Bovine, Rat, Mouse
- Pig, Human, Bovine, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- UniprotID# P41219
- UniprotID# P41219
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant full-length human Peripherin, expressed in and purified from E. coli
- Recombinant full-length human Peripherin, expressed in and purified from E. coli
- Form:
- Liquid
- Liquid
- Molecular weight:
- 57 kDa
- 57 kDa
- Storage buffer:
- Supplied as an aliquot of serum with 5 mM sodium azide
- Supplied as an aliquot of serum with 5 mM sodium azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
- Tested applications:
- ICC/IF, IHC
- ICC/IF, IHC
Specifications
Frequently Bought Together