Anti-Caspase-6 p18 Rabbit Polyclonal Antibody
ANTIA94575-100
New Product
- Antibody type:Primary
- Antigen name:Apoptotic protease Mch-2
- Antigen symbol:Caspase-6 p18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P55212
- Antigen synonyms:Apoptotic protease MCH2|CASP6|Caspase-6 subunit p11|Caspase 6|CASP-6|Caspase-6 p18|Caspase 6, apoptosis related cysteine peptidase|Caspase 6 apoptosis related cysteine protease|CASP6_HUMAN
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:33 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human Caspase 6 around the phosphorylation site of Ser257 (amino acids 223-272)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to Caspase 6 (phospho Ser257) for WB, IHC, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IHC, IF, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:40000
Type: Primary
Antigen: Caspase-6 p18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat