Anti-alpha 1 Fetoprotein Mouse Recombinant Antibody [clone: rAFP/7257]
Catalog # ANTIA314454-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Afp
- Antigen symbol:alpha 1 Fetoprotein
- Clonality:Recombinant
- Clone:rAFP/7257
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P02771
- Antigen synonyms:Alpha-fetoprotein|Alpha-fetoglobulin|Alpha fetoglobulin|AFPD|Alpha-1-fetoprotein|Alpha fetoprotein|Alpha fetoprotein precursor|alpha-fetoprotein, Hereditary persistence of, included|alpha 1 Fetoprotein
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:70 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human full-length AFP protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rAFP/7257] antibody to alpha 1 Fetoprotein for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: alpha 1 Fetoprotein
Clonality: Recombinant
Clone: rAFP/7257
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human