Anti-ALDH1A1 Mouse Recombinant Antibody [clone: rALDH1A1/7285]
Catalog # ANTIA314438-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Acetaldehyde dehydrogenase 1
- Antigen symbol:ALDH1A1
- Clonality:Recombinant
- Clone:rALDH1A1/7285
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P00352
- Antigen synonyms:AHD2|AL1A1_HUMAN|Aldehyde dehydrogenase 1 soluble|Aldehyde dehydrogenase|Aldehyde dehydrogenase cytosolic|ALDC|Aldehyde dehydrogenase family 1 member A1|Aldehyde dehydrogenase 1A1|Aldehyde dehydrogenase 1 family member A1
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:56 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 315-434) of human ALDH1A1 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rALDH1A1/7285] antibody to ALDH1A1 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: ALDH1A1
Clonality: Recombinant
Clone: rALDH1A1/7285
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human