Anti-RAB3IP Rabbit Polyclonal Antibody
ANTIA96624-100
New Product
- Antibody type:Primary
- Antigen name:Gtpat12
- Antigen symbol:RAB3IP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q96QF0
- Antigen synonyms:RAB3A interacting protein (rabin3)|RABIN3|Rab-3A-interacting protein|RAB3A interacting protein|RAB3A-interacting protein|Rabin-3|Rabin 3|RAB3IP|RAB3I_HUMAN
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:53 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human RAB3IP (amino acids 257-306)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to RAB3IP for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:40000
Type: Primary
Antigen: RAB3IP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat