Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Nucleophosmin Mouse Monoclonal Antibody [clone: ABT-NPM]
Anti-Nucleophosmin Mouse Monoclonal Antibody [clone: ABT-NPM]
Supplier:  ANTIBODIES.COM
New Product
Certificates
undefined
Anti-Nucleophosmin Mouse Monoclonal Antibody [clone: ABT-NPM]
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Catalog No:
  • ANTIA96294-100
  • ANTIA96294-50
  • Antigen symbol:
  • Nucleophosmin
  • Nucleophosmin
  • Antigen name:
  • B23
  • B23
  • Antigen synonyms:
  • NPM|MGC104254|NO38|NPM_HUMAN|Nucleophosmin|Nucleolar protein NO38|Nucleophosmin (nucleolar phosphoprotein B23 numatrin)|Nucleolar phosphoprotein B23|NPM1
  • NPM|MGC104254|NO38|NPM_HUMAN|Nucleophosmin|Nucleolar protein NO38|Nucleophosmin (nucleolar phosphoprotein B23 numatrin)|Nucleolar phosphoprotein B23|NPM1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • ABT-NPM
  • ABT-NPM
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • UniprotID# P06748
  • UniprotID# P06748
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide derived from human Nucleophosmin
  • Synthetic peptide derived from human Nucleophosmin
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Immunogen affinity purified
  • Immunogen affinity purified
  • Storage buffer:
  • Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
  • Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
  • Shipping temperature:
  • Shipped on blue ice at +4 °C
  • Shipped on blue ice at +4 °C
  • Tested applications:
  • IHC
  • IHC

Specifications

Frequently Bought Together