Anti-CD45R Rat Monoclonal Antibody [Clone: RA3-6B2] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA85840-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:B220
- Antigen symbol:CD45R
- Clonality:Monoclonal
- Clone:RA3-6B2
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Human,Mouse,Cat
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:CD45R|CD45|Glycoprotein|GP180|L CA|Leukocyte common antigen|L-CA|LCA|CD45 antigen
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Concentration:0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Abelson murine leukemia virus-induced pre-B tumor cells
- Purification:This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rat monoclonal [RA3-6B2] antibody to CD45R (FITC) for flow cytometry with samples derived from human, mouse and feline.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: CD45R
Clonality: Monoclonal
Clone: RA3-6B2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Cat