Anti-BAG3 Goat Polyclonal Antibody
Catalog # ANTIA83851-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:BAG 3
- Antigen symbol:BAG3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:BAG family molecular chaperone regulator 3|BCL2 associated athanogene 3|BAG3_HUMAN|Bcl 2 binding protein|BAG-3|Bcl-2-associated athanogene 3|BCL2 binding athanogene 3|Bag3|Bcl-2-binding protein Bis
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-SSMTDTPGNPAAP
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human BAG3 (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to BAG3 for ELISA and WB with samples derived from human.
- Validated applications: ELISA, W
Type: Primary
Antigen: BAG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human