Anti-KIF4A Goat Polyclonal Antibody
ANTIA83849-100
New Product
- Antibody type:Primary
- Antigen name:Chromokinesin
- Antigen symbol:KIF4A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:Chromosome-associated kinesin KIF4A|FLJ12655|Chromokinesin-A|FLJ14204|FLJ12530|KIF4|HSA271784|Chromosome associated kinesin KIF4A|FLJ20631
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:SFFSGCSPIEEEAH
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human KIF4A (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Goat polyclonal antibody to KIF4A for ELISA, WB, IF and flow cytometry with samples derived from human and mouse.
- Validated applications: ELISA, WB, IF, Flow Cytometr
Type: Primary
Antigen: KIF4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse