Order Entry
United Kingdom
ContactUsLinkComponent
Anti-CELF-6 Rabbit Polyclonal Antibody
Anti-CELF-6 Rabbit Polyclonal Antibody
Catalog # ANTIA306570-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-CELF-6 Rabbit Polyclonal Antibody
Catalog # ANTIA306570-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    6330569O16Rik
  • Antigen symbol:
    CELF-6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q96J87
  • Antigen synonyms:
    Bruno-like protein 6|bruno like 6, RNA binding protein|CELF6_HUMAN|CELF-6|CUG-BP- and ETR-3-like factor 6|Celf6|BRUNOL6|Bruno like protein 6|Bruno like 6, RNA binding protein (Drosophila)
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% Thiomersal
  • Molecular weight:
    51 kDa
  • Sequence:
    LGAFHPAPLPLGACGAYTTAILQHQAALLAAAQGPGLGPVAAVAAQMQHVAAFSLVAAPLLPAAAANSPPGSGPGTLPGLPAPIGVNGFGPLTPQTNGQPGSDTLYNNGLSPYPAQSPGVADPLQQAYAGMHHYAAAYPSAYAPVSTAFPQ
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 230-380 of human CELF6 (NP_443072.3).
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to CELF-6 for WB with samples derived from Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: CELF-6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat