Anti-Wilms Tumor Protein Mouse Monoclonal Antibody [clone: WT1/7452]
ANTIA317174-100
New Product
- Antibody type:Primary
- Antigen name:AWT1
- Antigen symbol:Wilms Tumor Protein
- Clonality:Monoclonal
- Clone:WT1/7452
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P19544
- Antigen synonyms:Wilms Tumor Protein|WAGR|FWT1|NPHS4|Wilms' tumor protein|Wilms Tumor|GUD|Wilms' tumor gene|Wilms tumor 1
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without Sodium azide and carrier free
- Molecular weight:49 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 150-350) of human WT1 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [WT1/7452] antibody to Wilms Tumor Protein for ELISA and IHC-P with samples derived from human.
- Validated applications: ELISA, IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Wilms Tumor Protein
Clonality: Monoclonal
Clone: WT1/7452
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human