Anti-Transglutaminase 6 Goat Polyclonal Antibody
ANTIA82747-100
New Product
- Antibody type:Primary
- Antigen name:Protein-glutamine gamma-glutamyltransferase 6
- Antigen symbol:Transglutaminase 6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:TGM3L_HUMAN|TGase-6|TGM6|Transglutaminase 6|TGase-3-like|TG6|TGase Y|TGY|Transglutaminase Y
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:C-DKYVDSYGRTLED
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Mouse Tgm6 (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Goat polyclonal antibody to Transglutaminase 6 for ELISA and WB with samples derived from rat.
- Validated applications: ELISA, WB
Type: Primary
Antigen: Transglutaminase 6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat