Order Entry
United Kingdom
ContactUsLinkComponent
Anti-CD30 Rabbit Recombinant Antibody [clone: DM104]
Anti-CD30 Rabbit Recombinant Antibody [clone: DM104]
Supplier:  ANTIBODIES.COM
New Product
Certificates
undefined
Anti-CD30 Rabbit Recombinant Antibody [clone: DM104]
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Catalog No:
  • ANTIA317355-500
  • ANTIA317355-100
  • Antigen symbol:
  • CD30
  • CD30
  • Antigen name:
  • CD 30
  • CD 30
  • Antigen synonyms:
  • CD30|KI1|Cytokine receptor CD30|KI 1|Ki-1 antigen|D1S166E|KI 1 antigen|CD30L receptor|CD30 antigen
  • CD30|KI1|Cytokine receptor CD30|KI 1|Ki-1 antigen|D1S166E|KI 1 antigen|CD30L receptor|CD30 antigen
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Recombinant
  • Recombinant
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • DM104
  • DM104
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • UniprotID# P28908
  • UniprotID# P28908
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Form:
  • Lyophilised
  • Lyophilised
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Concentration:
  • Reconstitution dependent
  • Reconstitution dependent
  • Storage buffer:
  • Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
  • Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
  • Storage temperature:
  • Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze / thaw cycles.
  • Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze / thaw cycles.
  • Shipping temperature:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C

Specifications

Frequently Bought Together