Anti-Perforin Mouse Monoclonal Antibody [clone: PRF1/2468]
ANTIA252892-100
New Product
- Antibody type:Primary
- Antigen name:Cytolysin
- Antigen symbol:Perforin
- Clonality:Monoclonal
- Clone:PRF1/2468
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P14222
- Antigen synonyms:Perforin-1|Perforin|P1|FLH2|Lymphocyte pore-forming protein|HPLH2|perforin 1 (pore forming protein)|PERF_HUMAN|Perforin 1
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:75 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 413 - 552, of human Perforin protein. The exact sequence is proprietary
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [PRF1/2468] antibody to Perforin for ELISA with samples derived from Human.
- Validated applications: ELISA
Type: Primary
Antigen: Perforin
Clonality: Monoclonal
Clone: PRF1/2468
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human