Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Glyoxalase 1 Mouse Monoclonal Antibody [clone: CPTC-GLO1-1]
Anti-Glyoxalase 1 Mouse Monoclonal Antibody [clone: CPTC-GLO1-1]
Catalog # ANTIA251921-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Glyoxalase 1 Mouse Monoclonal Antibody [clone: CPTC-GLO1-1]
Catalog # ANTIA251921-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Glyoxalase 1
  • Clonality:
    Monoclonal
  • Clone:
    CPTC-GLO1-1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q04760
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    24 - 26 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human GLO1 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [CPTC-GLO1-1] antibody to Glyoxalase 1 for WB and IHC-P with samples derived from Human.

  • Validated applications: WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Glyoxalase 1
Clonality: Monoclonal
Clone: CPTC-GLO1-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human