Anti-Annexin V Mouse Monoclonal Antibody [Clone: 6A12]
Supplier: ANTIBODIES.COM
New Product
Certificates
About this item
Mouse monoclonal [6A12] antibody to Annexin V for WB and ICC/IF with samples derived from Human and Monkey.
- Validated applications: WB, ICC/IF
- Recommended dilutions: WB: 1:2000 to 1:5000, ICC/IF: 1:1000 to 1:2000
Type: Primary
Antigen: Annexin V
Clonality: Monoclonal
Clone: 6A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Specifications
- Catalog No:
- ANTIA270559-50
- ANTIA270559-100
- Antigen symbol:
- Annexin V
- Annexin V
- Antigen name:
- Anchorin CII
- Anchorin CII
- Antigen synonyms:
- Calphobindin I|ANXA5|Annexin-5|Annexin V|Annexin 5|ANXA5_HUMAN|ANX5|ANX A5|Annexin A5
- Calphobindin I|ANXA5|Annexin-5|Annexin V|Annexin 5|ANXA5_HUMAN|ANX5|ANX A5|Annexin A5
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 6A12
- 6A12
- Reactivity:
- Monkey, Human
- Monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P08758
- UniprotID# P08758
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant full-length human Annexin A5, expressed in and purified from E. coli.
- Recombinant full-length human Annexin A5, expressed in and purified from E. coli.
- Form:
- Liquid
- Liquid
- Molecular weight:
- 35 kDa
- 35 kDa
- Purification:
- Immunogen affinity purification
- Immunogen affinity purification
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- Supplied in Phosphate Buffered Saline with 50% Glycerol and 5mM Sodium Azide
- Supplied in Phosphate Buffered Saline with 50% Glycerol and 5mM Sodium Azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
- Tested applications:
- ICC/IF
- ICC/IF
Specifications
Frequently Bought Together