Anti-beta Catenin Mouse Monoclonal Antibody [clone: 15B8]
Catalog # ANTIA251476-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:beta Catenin
- Antigen symbol:beta Catenin
- Clonality:Monoclonal
- Clone:15B8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Chicken,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P35222
- Antigen synonyms:CTNB1_HUMAN|CHBCAT|CATNB|Catenin beta 1|Catenin (cadherin associated protein), beta 1, 88kDa|Beta-catenin|Catenin beta-1|CTNNB|Cadherin associated protein
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:92 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant chicken beta Catenin
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [15B8] antibody to beta Catenin for Flow Cytometry, IF, WB and IHC-P with samples derived from Human, Mouse, Rat, Bovine, Canine and Chicken.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: beta Catenin
Clonality: Monoclonal
Clone: 15B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Bovine, Dog, Chicken