Order Entry
United Kingdom
ContactUsLinkComponent
Anti-CD27 Mouse Monoclonal Antibody [clone: LPFS2/4177]
Anti-CD27 Mouse Monoclonal Antibody [clone: LPFS2/4177]
Catalog # ANTIA250621-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-CD27 Mouse Monoclonal Antibody [clone: LPFS2/4177]
Catalog # ANTIA250621-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD 27
  • Antigen symbol:
    CD27
  • Clonality:
    Monoclonal
  • Clone:
    LPFS2/4177
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P26842
  • Antigen synonyms:
    CD27_HUMAN|S152|CD27 antigen|CD27 molecule|CD27L receptor|MGC20393|OTTHUMP00000238557|CD27|LPFS2
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    120 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 28 - 170, of human CD27 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [LPFS2/4177] antibody to CD27 for IHC-P with samples derived from Human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: LPFS2/4177
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human