Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Endothelin 1 Rabbit Polyclonal Antibody
Anti-Endothelin 1 Rabbit Polyclonal Antibody
  ANTIA12679-100
New Product
undefined
Anti-Endothelin 1 Rabbit Polyclonal Antibody
  ANTIA12679-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    ARCND3
  • Antigen symbol:
    Endothelin 1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P05305
  • Antigen synonyms:
    EDN1_HUMAN|ET1|ET-1|PPET1|EDN1|Preproendothelin 1|HDLCQ7|Big endothelin-1|Endothelin 1
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% Glycerol and 0,05% Proclin 300
  • Molecular weight:
    26 kDa
  • Sequence:
    APETAVLGAELSAVGENGGEKPTPSPPWRLRRSKRCSCSSLMDKECVYFCHLDIIWVNTPEHVVPYGLGSPRSKRALENLLPTKATDRENRCQCASQKDKKCWNFCQAGKELRAEDIMEKDWNNHKKGKDCSKLGKKCIYQQLVRGRKIRRSSEEHLRQTRSETMRNSVKSSFHDPKLKGKPSRERYVTHNRAHW
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 18-212 of human Endothelin 1 (NP_001946.3)
  • Tested applications:
    ICC/IF
  • Purification:
    Affinity purification.
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to Endothelin 1 for WB and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, ICC/IF
  • Recommended dilutions: WB: 1:500 to 1:1000, ICC/IF: 1:50 to 1:20

Type: Primary
Antigen: Endothelin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat